Learn More
BRAND™ PD-Tip™ Disposable Pipet Tips: Adapter
Accessory for BIO-CERT™ PD-Tip™ syringe tips.
182.00 DKK
Specifications
Volume (Metric) | 50 mL |
---|---|
Autoclavable | Autoclavable |
Disposable | Single-use |
Product Line | PD-Tip |
Material | Polypropylene |
Product Code | Brand | Sterility | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Packaging | Price | Quantity & Availability | |||||
10291641
|
BRAND™
702398 |
Non-sterile | Bulk packed |
182.00 DKK
Pack of 10 |
In Stock
Log in to see stock. ![]() |
|||||
Description
Adapter. As supplied with 25 and 50 mL PD-Tip Disposable Tips. Polypropylene. Autoclavable. BIO-CERT™ sterile adapters are DNA-, RNase-, ATP- and endotoxin-free, and individually wrapped.Specifications
50 mL | |
Single-use | |
Polypropylene |
Autoclavable | |
PD-Tip |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ PD-Tip™ Disposable Pipet Tips: Adapter