Learn More
Paraformaldehyde, 96%, extra pure
Paraformaldehyde, 96%, CH2O, CAS Number-30525-89-4, paraformaldehyde, paraform, oxymethylene, methylene oxide, methyl aldehyde, methanal, formol, formalin, formic aldehyde, oxomethane, 500g, CHEBI:16842, White, 260 deg.C, 1.03 (Air - 1.0), 71 deg.C, Extra Pure, Warning | CAS: 30525-89-4 | CH2O | 30.026 g/mol
226.00 DKK - 3305.00 DKK
Chemical Identifiers
CAS | 30525-89-4 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.026 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10351641
|
Thermo Scientific Acros
416780250 |
25 g | Plastic bottle |
226.00 DKK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10424131
|
Thermo Scientific Acros
416785000 |
500 g | Plastic Bottle |
347.00 DKK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10342243
|
Thermo Scientific
416780010 |
1 kg | Plastic bottle |
563.00 DKK
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10443361
|
Thermo Scientific Acros
416780030 |
3 kg | Plastic bottle |
1225.00 DKK
3kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924801
|
Thermo Scientific Acros
416780100 |
10 kg | Plastic Bucket |
3305.00 DKK
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
30525-89-4 | |
30.026 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
CH2O | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
Specifications
30525-89-4 | |
120.0°C to 170.0°C | |
1.03 (Air = 1.0) | |
71°C | |
96% | |
(CH2O)n | |
Solubility in water: sparingly soluble. Other solubilities: the higher polymers are insoluble in water, insoluble in alcohol and ether | |
C=O | |
30.026 | |
CHEBI:16842 | |
96% | |
Beads or Powder |
260°C | |
White | |
4.0 to 5.5 (10% suspension) | |
Authentic | |
CH2O | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
1.2mmHg at 25°C | |
Extra Pure | |
Paraformaldehyde, Extra Pure |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Suspected of causing cancer.
May cause an allergic skin reaction.
Harmful if inhaled.
Flammable in liquid state, upper limit 73%, lower limit 7.0%
Emergency Overview
Flammable in liquid state, upper limit 73%, lower limit 7.0%. Harmful by inhalation and if swallowed. Irritating to eyes, respiratory system and skin. Possible cancer hazard. May cause cancer based on animal data. May cause pulmonary edema.
Warning
- Paraformaldehyde
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Carcinogenicity Category 2
- Skin sensitiser Category 1
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H317-May cause an allergic skin reaction.
- H332-Harmful if inhaled.
- H318-Causes serious eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.