missing translation for 'onlineSavingsMsg'
Få mere at vide

Oxytetracycline, 96%, Thermo Scientific™

Oxytetracycline, CAS # 79-57-2, shows antibacterial, anti-inflammatory, and anti-proliferative activities. It can also inhibit mitochondrial protein synthesis.

408.65 DKK - 3544.83 DKK

Kemiske identifikatorer

CAS 79-57-2
Molekylær formel C22H26N2Na2O11
Molekylvægt (g/mol) 540.43
MDL nummer MFCD00003700
InChI nøgle OLRBUPVCLOPDOT-GETZKYKJSA-M
PubChem CID 54675779
IUPAC navn disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate
SMIL O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
Se flere specifikationer

Produkter 3
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
15434809
Dokumentvisning
Thermo Scientific Alfa Aesar
J62427.09
10g
ONLINE EXCLUSIVE PRICE
454.48 DKK
408.65 DKK / 10g
Spare 45.83 DKK
10% Slukket
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
15444809
Dokumentvisning
Thermo Scientific Alfa Aesar
J62427.18
50g
ONLINE EXCLUSIVE PRICE
1649.28 DKK
1484.65 DKK / 50g
Spare 164.63 DKK
9% Slukket
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
15454809
Dokumentvisning
Thermo Scientific Alfa Aesar
J62427.22
100g
3544.83 DKK
100g
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

  • Oxytetracycline is a compound with broad-spectrum antibacterial activity. Furthermore, it also shows anti-inflammatory and anti-proliferative activities
  • It can inhibit mitochondrial protein synthesis in both gram-positive and gram-negative bacteria

Application

  • Oxytetracycline is active against several species including Mycoplasma pneumonia, Pasteurella pestis, Escherichia coli, Haemophilus influenza, and Diplococcus pneumonia
  • In laboratory experiments, oxytetracycline can be used to evaluate the acquisition of oxytetracycline-resistance genes, which are associated with the development of antibiotic resistance
  • It can reduce cytochrome c activity, causing cell growth arrest
  • It has been shown to reduce murine bronchial inflammation
  • Oxytetracycline exerts its anti-inflammatory activity by decreasing the levels of IL-4, IL-5, IL-13 immune cells, and the Ovalbumin-specific IgE
  • Oxytetracycline can be used to inhibit NF-κB activation via phosphorylation and degradation of IκB-α
Tekniske data

Kemiske identifikatorer

79-57-2
540.43
OLRBUPVCLOPDOT-GETZKYKJSA-M
disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate
C22H26N2Na2O11
MFCD00003700
54675779
O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O

Tekniske data

Oxytetracycline
Tan to Yellow
MFCD00003700
Sparingly soluble in aqueous solution at 0.6 mg/mL
O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
540.43
460.43
79-57-2
C22H26N2Na2O11
14,6976
OLRBUPVCLOPDOT-GETZKYKJSA-M
disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate
54675779
≥98%
Videoer
Sikkerhed og håndtering

Sikkerhed og håndtering

P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P363-P501c

H302-H312-H319-H332-H360

missing translation for 'einecsNumber' : 201-212-8

missing translation for 'rtecsNumber' : QI7875000

missing translation for 'tsca' : Yes

Anbefalet opbevaring : Ambient temperatures

SDS
Dokumenter

Dokumenter

Produktcertifikater

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.