Få mere at vide
Oxytetracycline, 96%, Thermo Scientific™
Oxytetracycline, CAS # 79-57-2, shows antibacterial, anti-inflammatory, and anti-proliferative activities. It can also inhibit mitochondrial protein synthesis.
Brand: Thermo Scientific Alfa Aesar J62427.22
| Mængde | 100g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
- Oxytetracycline is a compound with broad-spectrum antibacterial activity. Furthermore, it also shows anti-inflammatory and anti-proliferative activities
- It can inhibit mitochondrial protein synthesis in both gram-positive and gram-negative bacteria
Application
- Oxytetracycline is active against several species including Mycoplasma pneumonia, Pasteurella pestis, Escherichia coli, Haemophilus influenza, and Diplococcus pneumonia
- In laboratory experiments, oxytetracycline can be used to evaluate the acquisition of oxytetracycline-resistance genes, which are associated with the development of antibiotic resistance
- It can reduce cytochrome c activity, causing cell growth arrest
- It has been shown to reduce murine bronchial inflammation
- Oxytetracycline exerts its anti-inflammatory activity by decreasing the levels of IL-4, IL-5, IL-13 immune cells, and the Ovalbumin-specific IgE
- Oxytetracycline can be used to inhibit NF-κB activation via phosphorylation and degradation of IκB-α
Kemiske identifikatorer
| 79-57-2 | |
| 540.43 | |
| OLRBUPVCLOPDOT-GETZKYKJSA-M | |
| disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate |
| C22H26N2Na2O11 | |
| MFCD00003700 | |
| 54675779 | |
| O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O |
Tekniske data
| Oxytetracycline | |
| Tan to Yellow | |
| C22H26N2Na2O11 | |
| 14,6976 | |
| OLRBUPVCLOPDOT-GETZKYKJSA-M | |
| disodium (6aS,8Z,10S,10aR,11S,11aR,12S)-8-[amino(hydroxy)methylidene]-10-(dimethylamino)-6a,11,12-trihydroxy-12-methyl-5,6,7,9-tetraoxo-5,5a,6,6a,7,8,9,10,10a,11,11a,12-dodecahydrotetracen-5a-id-4-olate dihydrate | |
| 54675779 | |
| ≥98% |
| 79-57-2 | |
| 100g | |
| MFCD00003700 | |
| Sparingly soluble in aqueous solution at 0.6 mg/mL | |
| O.O.[Na+].[Na+].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3[C-](C(=O)C4=C([O-])C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O | |
| 540.43 | |
| 460.43 |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.