Learn More
Invitrogen™ Human OXSR1, GST Tag Recombinant Protein
Recombinant Protein
4245.00 DKK - 61400.00 DKK
Specifications
Accession Number | O95747 |
---|---|
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Gene ID (Entrez) | 9943 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15910313
|
Invitrogen™
A30983 |
10 μg |
4245.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17138755
|
Invitrogen™
A32901 |
100 μg |
19780.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17108805
|
Invitrogen™
A33386 |
1 mg |
61400.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This OXSR1 recombinant human protein (full length) was expressed in insect cells. OXSR1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant
OXSR1 serine/threonine kinase or (OSR1) is activated by osmotic stresses and regulates downstream protein kinases such as PAK1 in order to establish the sensor/signaling modules that initiate the cellular response to environmental stress. OXSR1 can be phosphorylated by ATM or ATR during the DNA damage process. It regulates downstream kinases in response to environmental stress, and may play a role in regulating the actin cytoskeleton.Specifications
O95747 | |
Kinase Assay | |
9943 | |
Human OXSR1, GST Tag | |
OXSR1 | |
Unconjugated | |
Recombinant | |
Full length | |
Dry Ice | |
Full Length, Recombinant |
See Label | |
50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 | |
84.3 kDa | |
2210022N24Rik; 2810422B09Rik; AI462649; AW209236; C6orf182; CEP57R; KIAA1101; mKIAA1101; Osr1; OTTHUMP00000209062; oxidative stress responsive 1; oxidative stress responsive kinase 1; oxidative stress-responsive 1 protein; oxidative-stress responsive 1; OXSR1; Serine/threonine-protein kinase OSR1 | |
STE20 Kinase Family | |
Human | |
GST-tag | |
-80°C, Avoid Freeze/Thaw Cycles | |
Insect cells | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.