Få mere at vide
Invitrogen™ Human OXSR1, GST Tag Recombinant Protein
Beskrivelse
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This OXSR1 recombinant human protein (full length) was expressed in insect cells. OXSR1 recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen™ Eu Kinase Binding assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Full Length, Recombinant

Tekniske data
Tekniske data
Adgangsnummer | O95747 |
Koncentration | See Label |
Til brug med (applikation) | Kinase Assay |
Formulering | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Gen-id (Entrez) | 9943 |
Molekylvægt (g/mol) | 84.3 kDa |
Navn | Human OXSR1, GST Tag |
Mængde | 100 μg |
Gene Alias | 2210022N24Rik; 2810422B09Rik; AI462649; AW209236; C6orf182; CEP57R; KIAA1101; mKIAA1101; Osr1; OTTHUMP00000209062; oxidative stress responsive 1; oxidative stress responsive kinase 1; oxidative stress-responsive 1 protein; oxidative-stress responsive 1; OXSR1; Serine/threonine-protein kinase OSR1 |
Fælles navn | OXSR1 |
Vis mere |
Dokumenter
Product Information
Manuals
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.