missing translation for 'onlineSavingsMsg'
Learn More

Oxfendazole, Thermo Scientific™

205.00 DKK - 665.00 DKK

Chemical Identifiers

CAS 53716-50-0
Molecular Formula C15H13N3O3S
Molecular Weight (g/mol) 315.35
MDL Number 00801063
InChI Key BEZZFPOZAYTVHN-UHFFFAOYNA-N
IUPAC Name methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17505585
View Documents
Thermo Scientific Acros
469210010
1 g
205.00 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
17515585
View Documents
Thermo Scientific Acros
469210050
5 g
665.00 DKK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

53716-50-0
315.35
BEZZFPOZAYTVHN-UHFFFAOYNA-N
COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1
C15H13N3O3S
00801063
methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate

Specifications

Glass Bottle
Available
C15H13N3O3S
White to Light Yellow to Beige
BEZZFPOZAYTVHN-UHFFFAOYNA-N
methyl N-[6-(benzenesulfinyl)-1H-1,3-benzodiazol-2-yl]carbamate
Oxfendazole
53716-50-0
Powder/Crystals
00801063
13-7004
COC(=O)NC1=NC2=CC=C(C=C2N1)S(=O)C1=CC=CC=C1
315.35
Videos
Safety and Handling

Safety and Handling

Exclamation mark

missing translation for 'einecsNumber' : 258-714-5

missing translation for 'rtecsNumber' : FD0808000

Recommended Storage : Normal conditions

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Oxfendazole, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.