Learn More
Hughes Outdoor Self-Draining Safety Shower
Floor mounted self-draining drench shower with galvanized mild steel pipework.
7724.20 DKK - 8292.65 DKK
Product Code | Brand | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | |||||
17752358
|
Hughes
HSHSSD18GS |
Stainless Steel |
8292.65 DKK
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
17762358
|
Hughes
HSHSSD18GV |
Galvanized Steel |
7724.20 DKK
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
Description
- This ANSI compliant self-draining emergency safety shower is suitable for hot climates
- The design allows standing water to drain away from the standpipe to ensure the delivery of tepid water
- The valve is situated directly above the water inlet; the bleed pipe is fitted above this so standing water can drain into the ground preventing stagnant water overheating from solar radiation and removing the risk of further injury to the operator
- Available with galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Floor | |
Safety Shower |