Learn More
Macherey-Nagel™ Optima™ 210 GC Capillary Column
Brand: Macherey-Nagel™ 726877.30
This item is not returnable.
View return policy
Specifications
0.5 mm | |
0.32 mm | |
G1, G2, G38 | |
260°C to 280°C | |
30 m |
GC Capillary Column | |
Optima™ 210 | |
Gas Chromatography (GC) | |
0.25 ÎĽm | |
1 Each |
Product Content Correction
Product Title
Macherey-Nagel™ Optima™ 210 GC Capillary Column > Length: 30m; I.D.: 0.32mm; Film Thickness: 0.25um
Spot an opportunity for improvement?Share a Content Correction