Få mere at vide
Noradrenalone Hydrochloride, TRC
Artikelnummer. 30011992
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011992 500 mg
30007629 1 g
17918104 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011992 Leverandør Toronto Research Chemicals Leverandørnr. TRCN661035500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5090-29-9
Molekylær formel C8H10ClNO3
Molekylvægt (g/mol) 203.62
Synonym 2-Amino-3',4'-dihydroxyacetophenone Hydrochloride, 2-Amino-1-(3,4-dihydroxyphenyl)ethanone Hydrochloride
IUPAC navn 2-amino-1-(3,4-dihydroxyphenyl)ethanone;hydrochloride
SMIL Cl.O=C(C1=CC=C(O)C(O)=C1)CN
Tekniske data
Kemisk navn eller materiale Noradrenalone Hydrochloride
Mængde 500 mg
InChI formel InChI=1S/C8H9NO3.ClH/c9-4-8(12)5-1-2-6(10)7(11)3-5;/h1-3,10-11H,4,9H2;1H
Formel vægt 203.0349