Learn More
Hamilton™ Non-Filtered Low Retention Racked Pipette Tips
With surface walls completely free of occlusions and cavities so fluid retention is virtually eliminated.
Brand:  Hamilton™ 68007-21
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15355094
  • 860.00 DKK / Pack of 10
Estimated Shipment: 08-07-2024
to see stock.
Add to basket
Description
Description
  • Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
  • Available pre-sterilized or non-sterile
  • 96 tips to a rack
  • Delivering outstanding accuracy and precision, guaranteed
Specifications
Specifications
10 μL
Non-sterile
Virgin Polypropylene
Non-Filtered
12 racks; 1152 total tips