Learn More
Hamilton™ Non-Filtered Low Retention Racked Pipette Tips
With surface walls completely free of occlusions and cavities so fluid retention is virtually eliminated.
428.00 DKK - 860.00 DKK
Product Code | Brand | Volume (Metric) | Sterility | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Sterility | Quantity | Price | Quantity & Availability | |||
15325084
|
Hamilton™
68009-21 |
300 μL | Non-sterile | 10 racks; 960 total tips |
858.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
15325094
|
Hamilton™
68007-24 |
10 μL | Non-sterile | 10 racks; 960 total tips |
720.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
15355094
|
Hamilton™
68007-21 |
10 μL | Non-sterile | 12 racks; 1152 total tips |
860.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
15375074
|
Hamilton™
68010-22 |
1000 μL | Sterile | 6 racks; 576 total tips |
625.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
15385074
|
Hamilton™
68010-21 |
1000 μL | Non-sterile | 6 racks; 576 total tips |
428.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
15385084
|
Hamilton™
68008-22 |
200 μL | Sterile | 10 racks; 960 total tips |
800.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
15395084
|
Hamilton™
68008-21 |
200 μL | Non-sterile | 10 racks; 960 total tips |
720.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||
Description
- Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
- Available pre-sterilized or non-sterile
- 96 tips to a rack
- Delivering outstanding accuracy and precision, guaranteed
Specifications
Non-Filtered |