missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol

520.00 DKK - 4755.00 DKK

Chemical Identifiers

CAS 110-26-9
Molecular Formula C7H10N2O2
Molecular Weight (g/mol) 154.169
MDL Number MFCD00008625
InChI Key ZIUHHBKFKCYYJD-UHFFFAOYSA-N
Synonym MBA
PubChem CID 8041
IUPAC Name N-[(prop-2-enoylamino)methyl]prop-2-enamide
SMILES C=CC(=O)NCNC(=O)C=C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10226120
SDS View Documents
Thermo Scientific Chemicals
164790250
25 g Plastic Bottle
520.00 DKK
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10194030
SDS View Documents
Thermo Scientific Chemicals
164791000
100 g Plastic Bottle
1480.00 DKK
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10195533
SDS View Documents
Thermo Scientific Chemicals
164795000
500 g Plastic Bottle
4755.00 DKK
500g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.