Få mere at vide
(-)-Methylephedrine Hydrochloride, TRC
Artikelnummer. 17971895
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17971895 10 mg
17923935 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17971895 Leverandør Toronto Research Chemicals Leverandørnr. TRCM85050010MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 38455-90-2
Molekylær formel C11H17NOClH
Molekylvægt (g/mol) 215.7
Synonym (αR)-α-[(1S)-1-(Dimethylamino)ethyl]benzenemethanol hydrochloride (1:1), [R-(R*,S*)]-α-[1-(Dimethylamino)ethyl]benzenemethanol hydrochloride, (-)-Methylephedrine hydrochloride, L-erythro-Methylephedrine hydrochloride, Methy F, N-Methylephedrine hydrochloride, NSC 42289, L-N-Methylephedrine hydrochloride
IUPAC navn (1R,2S)-2-(dimethylamino)-1-phenylpropan-1-ol;hydrochloride
SMIL Cl.C[C@@H]([C@H](O)c1ccccc1)N(C)C
Tekniske data
Kemisk navn eller materiale (-)-Methylephedrine hydrochloride
Mængde 10 mg
InChI formel InChI=1S/C11H17NO.ClH/c1-9(12(2)3)11(13)10-7-5-4-6-8-10;/h4-9,11,13H,1-3H3;1H/t9-,11-;/m0./s1
Formel vægt 215.7197