Learn More
Magnesium Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
406.00 DKK - 840.00 DKK
Specifications
Solution Contains | H2O |
---|---|
Concentration | 1 g/L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12922145
![]() |
Fisher Chemical
A 025 100 |
100 mL |
406.00 DKK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12932145
![]() |
Fisher Chemical
A 025 250 |
250 mL |
576.00 DKK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12942145
![]() |
Fisher Chemical
A 025 500 |
500 mL |
840.00 DKK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
H2O | |
1 g/L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Magnesium Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™