Learn More
Magnesium Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™ Product Code.: 12932145
Magnesium Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™
BIOPHARMA
Product Code. 12932145 Shop All Fisher Chemical Products
250 mL, 250mL
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
Product Code. 12932145
Brand: Fisher Chemical A 025 250
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Solution Contains H2O
Concentration 1 g/L
Quantity 250 mL
Product Content Correction
Product Title
Magnesium Solution for Ion Chromatography, 1g/L, H2O Matrix, Fisher Chemical™ >