missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human KIT [V559D], GST Tag Recombinant Protein
Recombinant Protein
4245.00 DKK - 61400.00 DKK
Specifications
Accession Number | P10721 |
---|---|
Concentration | See Label |
For Use With (Application) | Kinase Assay |
Format | Kit |
Formulation | 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15920283
|
Invitrogen™
A30497 |
10 ÎĽg |
4245.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17128735
|
Invitrogen™
A32867 |
100 ÎĽg |
19780.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17148775
|
Invitrogen™
A33356 |
1 mg |
61400.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIT receptor tyrosine kinase is a proto-oncogene and a type 3 transmembrane receptor for MGF (mast cell growth factor, also known as stem cell factor). KIT was first identified as the cellular homolog of the feline sarcoma viral oncogene v-kit. KIT together with its ligand regulates growth and activation of a variety of hemopoietic and non-hemopoietic cells. Mutations in KIT are associated with gastrointestinal stromal tumors, mast cell disease, acute myelogenous leukemia, and piebaldism. Recently, deregulation of the KIT receptor TK by the prevalent activation loop mutation D816V has served as a focal point in therapeutic strategies aimed at curbing neoplastic mast cell growth.Specifications
P10721 | |
Kinase Assay | |
50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5 | |
75.3 kDa | |
KIT | |
PDGFR Kinase Family | |
Unconjugated | |
Recombinant | |
-80°C, Avoid Freeze/Thaw Cycles | |
Insect cells | |
Liquid |
See Label | |
Kit | |
3815 | |
Human KIT [V559D], GST Tag | |
KIT (V559D Mutant) | |
544-end | |
Human | |
GST-tag | |
Dry Ice | |
Recombinant, Active, Catalytic, Mutant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human KIT [V559D], GST Tag Recombinant Protein