Learn More
DWK Life Sciences Kimble™ Adapters for Round-Bottom Centrifuge Tubes
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
471.00 DKK - 725.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11819913
|
DWK Life Sciences
45550-15 |
15mL Round bottom centrifuge tube |
471.00 DKK
Pack of 2 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
11843825
|
DWK Life Sciences
45550-30_OLD |
30mL Round bottom centrifuge tube |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Required when centrifuging select Kimble high-strength round-bottom centrifuge tubes to 13,000xG in 50mL rotor
- Made of rubber
Specifications
Rubber |