Få mere at vide
Hydroxy Vardenafil, TRC
Artikelnummer. 30132618
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30132618 25 mg -
30011437 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30132618 Leverandør Toronto Research Chemicals Leverandørnr. TRCH99530025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 224785-98-2
Molekylær formel C23 H32 N6 O5 S
Molekylvægt (g/mol) 504.6
Synonym Imidazo[5,1-f][1,2,4]triazin-4(1H)-one, 2-[2-ethoxy-5-[[4-(2-hydroxyethyl)-1-piperazinyl]sulfonyl]phenyl]-5-methyl-7-propyl-, 1-Piperazineethanol, 4-[[3-(1,4-dihydro-5-methyl-4-oxo-7-propylimidazo[5,1-f][1,2,4]triazin-2-yl)-4-ethoxyphenyl]sulfonyl]- (9CI)
IUPAC navn 2-[2-ethoxy-5-[4-(2-hydroxyethyl)piperazin-1-yl]sulfonylphenyl]-5-methyl-7-propyl-3H-imidazo[5,1-f][1,2,4]triazin-4-one
SMIL CCCc1nc(C)c2C(=O)NC(=Nn12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(CCO)CC4
Tekniske data
Kemisk navn eller materiale Hydroxyvardenafil
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C23H32N6O5S/c1-4-6-20-24-16(3)21-23(31)25-22(26-29(20)21)18-15-17(7-8-19(18)34-5-2)35(32,33)28-11-9-27(10-12-28)13-14-30/h7-8,15,30H,4-6,9-14H2,1-3H3,(H,25,26,31)
Formel vægt 504.2155
Procent renhed >95%