missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Complement Factor I Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
1540.00 DKK - 3700.00 DKK
Tekniske data
| Antigen | Complement Factor I |
|---|---|
| Klon | 271203 |
| Koncentration | LYOPH |
| Fortynding | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applikationer | Western Blot, Immunoprecipitation |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
18786718
![]() |
R&D Systems
MAB3307-SP |
25 μg |
1540.00 DKK
25µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
18168810
![]() |
R&D Systems
MAB3307 |
100 μg |
3700.00 DKK
100µg |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Complement Factor I Monoclonal specifically detects Complement Factor I in Human samples. It is validated for Western Blot, Immunoprecipitation.Tekniske data
| Complement Factor I | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P05156 | |
| 3426 | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor I Lys19-Val583 Accession # P05156 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Complement Factor I in direct ELISAs and Western blots. |
| 271203 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AHUS3, C3b Inactivator, C3B/C4B inactivator, C3BINA, C3b-inactivator, CFI, complement component I, complement control protein factor I, complement factor I, complement factor I heavy chain, EC 3.4.21, EC 3.4.21.45, FIC3b-INA, I factor (complement), IF, KAF, Konglutinogen-activating factor, light chain of factor I | |
| CFI | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel