missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Complement Factor I Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB3307-SP
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Complement Factor I Monoclonal specifically detects Complement Factor I in Human samples. It is validated for Western Blot, Immunoprecipitation.
Tekniske data
| Complement Factor I | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P05156 | |
| CFI | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor I Lys19-Val583 Accession # P05156 | |
| 25 μg | |
| Primary | |
| Detects human Complement Factor I in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 271203 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AHUS3, C3b Inactivator, C3B/C4B inactivator, C3BINA, C3b-inactivator, CFI, complement component I, complement control protein factor I, complement factor I, complement factor I heavy chain, EC 3.4.21, EC 3.4.21.45, FIC3b-INA, I factor (complement), IF, KAF, Konglutinogen-activating factor, light chain of factor I | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3426 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion