missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Human Complement Factor I Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB3307
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Complement Factor I Monoclonal specifically detects Complement Factor I in Human samples. It is validated for Western Blot, Immunoprecipitation.
Tekniske data
| Complement Factor I | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P05156 | |
| CFI | |
| Mouse myeloma cell line NS0-derived recombinant human Complement Factor I Lys19-Val583 Accession # P05156 | |
| 100 μg | |
| Primary | |
| Detects human Complement Factor I in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 271203 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AHUS3, C3b Inactivator, C3B/C4B inactivator, C3BINA, C3b-inactivator, CFI, complement component I, complement control protein factor I, complement factor I, complement factor I heavy chain, EC 3.4.21, EC 3.4.21.45, FIC3b-INA, I factor (complement), IF, KAF, Konglutinogen-activating factor, light chain of factor I | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3426 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering