missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Hoechst 33342 analog 2 trihydrochloride

MedChemExpress Hoechst 33342 analog 2 trihydrochloride is a anglog of Hoechst 33342. Hoechst 33342 is a DNA minor groove binder used fluorochrome for visualizing cellular DNA.

Encompass_Preferred

4370.91 DKK - 18732.49 DKK

Specifications

Content And Storage 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Shipping Condition Room temperature
Solubility H2O : 20 mg/mL (30.31 mM; Need ultrasonic) ∣ DMSO : 4 mg/mL (6.06 mM; Need ultrasonic)
Molecular Weight (g/mol) 659.78 g/mol
CAS 155815-98-8
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16499257
Encompass_Preferred
View Documents
MedChemExpress
HY-15630A
5 mg
4370.91 DKK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16442787
Encompass_Preferred
View Documents
MedChemExpress
HY-15630A
50 mg
18732.49 DKK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16426708
Encompass_Preferred
View Documents
MedChemExpress
HY-15630A
10 mg
6244.17 DKK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
H2O : 20 mg/mL (30.31 mM; Need ultrasonic) ∣ DMSO : 4 mg/mL (6.06 mM; Need ultrasonic)
155815-98-8
Room temperature
659.78 g/mol
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
MedChemExpress Hoechst 33342 analog 2 trihydrochloride

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.