Learn More
MedChemExpress Hoechst 33342 analog 2 trihydrochloride
MedChemExpress Hoechst 33342 analog 2 trihydrochloride is a anglog of Hoechst 33342. Hoechst 33342 is a DNA minor groove binder used fluorochrome for visualizing cellular DNA.
Brand: MedChemExpress HY-15630A
Specifications
4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) | |
Room temperature | |
659.78 g/mol |
50 mg | |
H2O : 20 mg/mL (30.31 mM; Need ultrasonic) ∣ DMSO : 4 mg/mL (6.06 mM; Need ultrasonic) | |
155815-98-8 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.