Learn More
MedChemExpress Hoechst 33342 analog 2 trihydrochloride
MedChemExpress Hoechst 33342 analog 2 trihydrochloride is a anglog of Hoechst 33342. Hoechst 33342 is a DNA minor groove binder used fluorochrome for visualizing cellular DNA.
Brand:  MedChemExpress HY-15630A
Encompass_Preferred
 View more versions of this product
Product Code. 16426708
  • 6244.17 DKK / 10mg
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light)
Room temperature
659.78 g/mol
10 mg
H2O : 20 mg/mL (30.31 mM; Need ultrasonic) ∣ DMSO : 4 mg/mL (6.06 mM; Need ultrasonic)
155815-98-8
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
MedChemExpress Hoechst 33342 analog 2 trihydrochloride