Learn More
H-1152 dihydrochloride, MedChemExpress
MedChemExpress H-1152 dihydrochloride is a membrane-permeable and selective ROCK inhibitor, with a Ki value of 1.6 nM, and an IC50 value of 12 nM for ROCK2.

1873.25 DKK - 3021.37 DKK
Specifications
Chemical Name or Material | H-1152 dihydrochloride |
---|---|
CAS | 871543-07-6 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C16H23Cl2N3O2S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16459317
![]() |
Medchem Express
HY-15720A |
5 mg |
1873.25 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16456768
![]() |
Medchem Express
HY-15720A |
10 mg |
3021.37 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
H-1152 dihydrochloride is a membrane-permeable and selective ROCK inhibitor, with a Ki value of 1.6 nM, and an IC50 value of 12 nM for ROCK2.Specifications
H-1152 dihydrochloride | |
Light Yellow | |
C16H23Cl2N3O2S | |
CC1=CN=CC2=C1C(S(=O)(N3[C@@H](C)CNCCC3)=O)=CC=C2.[H]Cl.[H]Cl | |
392.34 | |
Research | |
Neuroscience-Neuromodulation |
871543-07-6 | |
Research | |
DMSO : 50 mg/mL (127.44 mM; Need ultrasonic) ∣H2O : 35.71 mg/mL (91.02 mM; Need ultrasonic) | |
392.34 | |
98.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.