missing translation for 'onlineSavingsMsg'
Learn More

Guanethidine sulfate, Thermo Scientific™

664.00 DKK - 2040.00 DKK

Chemical Identifiers

CAS 645-43-2
Molecular Formula C10H24N4O4S
Molecular Weight (g/mol) 296.39
MDL Number MFCD00035403
InChI Key YUFWAVFNITUSHI-UHFFFAOYSA-N
IUPAC Name N''-[2-(azocan-1-yl)ethyl]guanidine; sulfuric acid
SMILES OS(O)(=O)=O.NC(N)=NCCN1CCCCCCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17322491
View Documents
Thermo Scientific Acros
468622500
250 mg
664.00 DKK
250mg
Please to purchase this item. Need a web account? Register with us today!
17312491
View Documents
Thermo Scientific Acros
468620010
1 g
2040.00 DKK
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS07: Exclamation mark

missing translation for 'shelfLife' : 2 years

missing translation for 'einecsNumber' : 211-442-0

missing translation for 'rtecsNumber' : MF3150000

Recommended Storage : Normal conditions

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Guanethidine sulfate, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.