Learn More
Lenz Laborglasintrumente Flat-Bottom Flasks With Conical Ground Joint
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
630.00 DKK - 6985.00 DKK
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
10141471
|
Lenz Laborglasintrumente
03011937 |
100 mL | NS 19/26 | 64 mm |
1320.00 DKK
Pack of 10 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
10643501
|
Lenz Laborglasintrumente
03012937 |
100 mL | NS 29/32 | 64 mm |
630.00 DKK
Pack of 10 |
Estimated Shipment: 22-08-2025
Log in to see stock. |
||||
10433491
|
Lenz Laborglasintrumente
03012973 |
2 L | NS 29/32 | 166 mm |
6125.00 DKK
Pack of 10 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
10060211
|
Lenz Laborglasintrumente
03011949 |
250 mL | NS 19/26 | 85 mm |
2310.00 DKK
Pack of 10 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
10274141
|
Lenz Laborglasintrumente
03012473 |
2 L | NS 24/29 | 166 mm |
6985.00 DKK
Pack of 10 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
10460703
|
Lenz Laborglasintrumente
03012428 |
50 mL | NS 24/29 | 51 mm |
1085.00 DKK
Pack of 10 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
10500122
|
Lenz Laborglasintrumente
03012437 |
100 mL | NS 24/29 | 64 mm |
1260.00 DKK
Pack of 10 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
10694382
|
Lenz Laborglasintrumente
03011958 |
500 mL | NS 19/26 | 105 mm |
3435.00 DKK
Pack of 10 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
10767891
|
Lenz Laborglasintrumente
03012470 |
1 L | NS 24/29 | 131 mm |
4255.00 DKK
Pack of 10 |
Estimated Shipment: 10-09-2025
Log in to see stock. |
||||
Description
- According to DIN EN ISO 4797
- Wiith inscription label
- Material: DURAN™
Specifications
Conical ground joint and inscription label |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Flat-Bottom Flasks With Conical Ground Joint