Learn More
Thermo Scientific™ Fiberlite 1000mL Bottles
Thermo Scientific™ 1000 mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottles are available for Thermo Scientific™ Sorvall™ LYNX Superspeed and Bios A Large-capacity centrifuges
2204.49 DKK - 3066.79 DKK
Specifications
Capacity | 1000 mL |
---|---|
Capacity (Metric) | 1000 mL |
Capacity (English) | 33.81 oz. |
Color | Clear |
No. per Pack | Pack of 2 |
Product Code | Brand | Material | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | ||||||
15861392
|
Thermo Scientific™
010-1491 |
Polypropylene |
2204.49 DKK
Pack of 2 |
Estimated Shipment: 07-01-2026
Log in to see stock. |
||||||
15840134
|
Thermo Scientific™
010-1492 |
Polycarbonate |
3066.79 DKK
Pack of 2 |
Estimated Shipment: 25-08-2026
Log in to see stock. |
||||||
Description
Thermo Scientific™ 1000mL Fiberlite Bottles combine a Nalgene bottle body with newly designed Fiberlite Cap Closure. These bottles are available for Thermo Scientific™ Sorvall™ LYNX Superspeed and Bios A Large-capacity centrifuges
Features
• PPCO & PC
• Increased cycle life compared to previous Thermo Scientific bottles - 100 cycle life (caps + bottles)
• Certifications USP Class VI - Non-Cytotoxic
• Max speeds match rotors
• Ergonomic speed handle design allows much easier bottle handling and removal
Certifications
Certifications USP Class VI - Non-Cytotoxic
Specifications
1000 mL | |
33.81 oz. | |
Pack of 2 | |
USP Class VI Non-Cytotoxic | |
Round | |
Sealing Cap |
1000 mL | |
Clear | |
Fiberlite | |
No | |
Centrifuge Bottle | |
Yes |