Learn More
Ethyl pipecolinate, 98+% Product Code.: 10046353
Ethyl pipecolinate, 98+%
Product Code. p-3523445 Shop All Thermo Scientific Chemicals Products
5 g, Glass bottle, 5g
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. 10046353
Brand: Thermo Scientific Acros 189580050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 15862-72-3 | C8H16NO2 | 158.22 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15862-72-3
Molecular Formula C8H16NO2
Molecular Weight (g/mol) 158.22
MDL Number MFCD00005980
InChI Key SZIKRGHFZTYTIT-ZETCQYMHSA-O
Synonym ethyl pipecolinate, ethyl 2-piperidinecarboxylate, ethyl pipecolate, pipecolinic acid ethyl ester, pipecolic acid ethyl ester, ethylpiperidine-2-carboxylate, 2-piperidinecarboxylic acid ethyl ester, 2-piperidinecarboxylic acid, ethyl ester, ethylpipecolinate, pubchem7720Show MoreShow Less
PubChem CID 27517
IUPAC Name ethyl piperidine-2-carboxylate
SMILES CCOC(=O)[C@@H]1CCCC[NH2+]1
Specifications
Color Colorless to Yellow
Density 1.0060g/mL
Boiling Point 216°C to 217°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4550 to 1.457
Quantity 5 g
Beilstein 22, 7
Specific Gravity 1.006
Solubility Information Solubility in water: miscible
Formula Weight 157.21
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Ethyl pipecolinate
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0390789 Certificate TypeCertificate of Analysis Date26/05/2025 Product Code189580250,189580050
Lot NumberA0390789 Certificate TypeCertificate of Analysis Date23/11/2022 Product Code189580250,189580050
compliance-icons compliance-icons
Product Identifier
  • Ethyl pipecolinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Product Title
Ethyl pipecolinate, 98+% >