Få mere at vide
Ethyl 3-(3-(tert-Butyl)-4-hydroxy-5-methylphenyl)propanoate, TRC
Artikelnummer. 30180086
Skift visning
Mængde:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30180086 50 mg
30180146 250 mg
30180235 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30180086 Leverandør Toronto Research Chemicals Leverandørnr. TRCE17824050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 40200-35-9
Molekylær formel C16 H24 O3
Molekylvægt (g/mol) 264.36
Synonym Benzenepropanoic acid, 3-(1,1-dimethylethyl)-4-hydroxy-5-methyl-, ethyl ester, Ethyl 3-(3-methyl-5-tert-butyl-4-hydroxyphenyl)propionate, Ethyl 3-(3-tert-butyl-4-hydroxy-5-methylphenyl)propionate, Ethyl 3-(3'-methyl-5'-tert-butyl-4'-hydroxyphenyl)propionate, Ethyl β-(5-tert-butyl-4-hydroxy-3-methylphenyl)propionate
IUPAC navn ethyl 3-(3-tert-butyl-4-hydroxy-5-methylphenyl)propanoate
SMIL CCOC(=O)CCc1cc(C)c(O)c(c1)C(C)(C)C
Tekniske data
Kemisk navn eller materiale Ethyl 3-(3-tert-Butyl-4-hydroxy-5-methylphenyl)propionate
Mængde 50 mg
InChI formel InChI=1S/C16H24O3/c1-6-19-14(17)8-7-12-9-11(2)15(18)13(10-12)16(3,4)5/h9-10,18H,6-8H2,1-5H3
Formel vægt 264.1725