Learn More
Invitrogen™ eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
The Phytohemagglutinin-L (PHA-L) Solution (500X) is a ready to use solution of PHA-L in aqueous buffer.
678.00 DKK - 1802.00 DKK
Specifications
Product Line | eBioscience™ |
---|---|
Product Type | Phytohemagglutinin-L (PHA-L) Solution |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15546286
|
Invitrogen™
00-4977-03 |
4 x 100 ÎĽL |
1802.00 DKK
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15556286
|
Invitrogen™
00-4977-93 |
100 ÎĽL |
678.00 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- A ready to use solution of PHA-L in aqueous buffer
- PHA is a mixture of lectins isolated from the red kidney bean, Phaseolus vulgaris
- It is the fraction of PHA that exhibits strong leukoagglutinating and mitogenic activity, but very little erythroagglutinating activity
- This reagent is intended for use in in vitro activation of human and mouse leukocytes
Specifications
eBioscience™ | |
Phytohemagglutinin-L (PHA-L) Solution |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Invitrogen™ eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)