Få mere at vide
Diethyl ethylphenylmalonate, 96%, Thermo Scientific™
Artikelnummer. 11382137
Skift visning
Mængde:
5 g
25 g
Pakningsstørrelse:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11382137 5 g 5g
11392137 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11382137 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. L07998.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76-67-5
Molekylær formel C15H20O4
Molekylvægt (g/mol) 264.321
MDL nummer MFCD00040759
InChI nøgle PKRVDBARWFJWEB-UHFFFAOYSA-N
Synonym diethyl 2-ethyl-2-phenylmalonate, diethyl ethylphenylmalonate, diethyl ethyl phenyl malonate, propanedioic acid, ethylphenyl-, diethyl ester, ethylphenylmalonic acid diethyl ester, diethyl 2-ethyl-2-phenyl-propanedioate, propanedioic acid, 2-ethyl-2-phenyl-, 1,3-diethyl ester, 1,3-diethyl 2-ethyl-2-phenylpropanedioate, diethyl 2-ethyl-2-phenylpropane-1,3-dioate, malonic acid, ethylphenyl-, diethyl ester
PubChem CID 66450
IUPAC navn diethyl 2-ethyl-2-phenylpropanedioate
SMIL CCC(C1=CC=CC=C1)(C(=O)OCC)C(=O)OCC
Tekniske data
Kemisk navn eller materiale Diethyl ethylphenylmalonate
Tæthed 1.07
Kogepunkt 185°C (15 mmHg)
Flammepunkt >110°C (230°F)
Brydningsindeks 1.491
Mængde 5 g
Beilstein 668299
Formel vægt 264.32
Procent renhed 96%