Få mere at vide
Descarboxy 1-Hydroxy Ketorolac, TRC
Artikelnummer. 30008856
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008856 25 mg
30005399 50 mg
30004863 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008856 Leverandør Toronto Research Chemicals Leverandørnr. TRCD28911525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 154476-25-2
Molekylær formel C14 H13 N O2
Molekylvægt (g/mol) 227.26
Synonym Methanone, (2,3-dihydro-1-hydroxy-1H-pyrrolizin-5-yl)phenyl-, (2,3-Dihydro-1-hydroxy-1H-pyrrolizin-5-yl)phenylmethanone, (1RS)-5-Benzoyl-2,3-dihydro-1H-pyrrolizin-1-ol, Ketorolac 1-Hydroxy Analog, Descarboxy 1-Hydroxy Ketorolac
IUPAC navn (7-hydroxy-6,7-dihydro-5H-pyrrolizin-3-yl)-phenylmethanone
SMIL OC1CCn2c1ccc2C(=O)c3ccccc3
Tekniske data
Kemisk navn eller materiale 5-benzoyl-2,3-dihydro-1H-pyrrolizin-1-ol
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C14H13NO2/c16-13-8-9-15-11(13)6-7-12(15)14(17)10-4-2-1-3-5-10/h1-7,13,16H,8-9H2
Formel vægt 227.0946
Procent renhed >95%