Learn More
Thermo Scientific Chemicals D-(+)-Mannose, 99%
CAS: 3458-28-4 | C6H12O6
402.00 DKK - 4635.00 DKK
Chemical Identifiers
CAS | 3458-28-4 |
---|---|
MDL Number | MFCD00064122 |
Synonym | 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11449812
|
Thermo Scientific Chemicals
A10842.14 |
25 g |
402.00 DKK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459812
|
Thermo Scientific Chemicals
A10842.22 |
100 g |
1135.00 DKK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11469812
|
Thermo Scientific Chemicals
A10842.36 |
500 g |
4635.00 DKK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
D-mannose acts as an alternative energy source utilized by brain. It is used in the study of assessing the synthesis of amphiphilic glycopolymers and investigating early detection of bronchiolitis obliterans.It may be useful in the treatment of urinary tract infections (UTIs) and carbohydrate-deficient glycoprotein syndrome type 1b. It is also useful in the formation of glycan structure and glycosylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsD-mannose acts as an alternative energy source utilized by brain. It is used in the study of assessing the synthesis of amphiphilic glycopolymers and investigating early detection of bronchiolitis obliterans.It may be useful in the treatment of urinary tract infections (UTIs) and carbohydrate-deficient glycoprotein syndrome type 1b. It is also useful in the formation of glycan structure and glycosylation.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
3458-28-4 | |
MFCD00064122 | |
2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers |
Specifications
3458-28-4 | |
1.54 g/mL | |
C6H12O6 | |
1564373 | |
2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers | |
180.16 |
127°C to 133°C | |
99% | |
MFCD00064122 | |
Hygroscopic | |
Soluble in water. | |
D-(+)-Mannose |
Safety and Handling
missing translation for 'einecsNumber' : 222-392-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.