Learn More
Cyclosporin A, MedChemExpress
MedChemExpress Cyclosporin A (Cyclosporine A) is an immunosuppressant which binds to the cyclophilin and inhibits phosphatase activity of calcineurin with an IC50 of 5 nM. Cyclosporin A also inhibits CD11a/CD18 adhesion.

624.42 DKK - 1248.83 DKK
Specifications
Chemical Name or Material | Cyclosporin A |
---|---|
Physical Form | Solid |
CAS | 59865-13-3 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16471426
![]() |
Medchem Express
HY-B0579 |
200 mg |
876.19 DKK
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16438376
![]() |
Medchem Express
HY-B0579 |
100 mg |
624.42 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16462176
![]() |
Medchem Express
HY-B0579 |
500 mg |
1248.83 DKK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cyclosporin A (Cyclosporine A) is an immunosuppressant which binds to the cyclophilin and inhibits phosphatase activity of calcineurin with an IC50 of 5 nM. Cyclosporin A also inhibits CD11a/CD18 adhesion.Specifications
Cyclosporin A | |
59865-13-3 | |
Research | |
COVID-19-immunoregulation | |
DMSO : 62.5 mg/mL (51.97 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (41.58 mM; Need ultrasonic) | |
1202.61 | |
98.0% |
Solid | |
White | |
C62H111N11O12 | |
Cyclosporine A Ciclosporin A CsA | |
C/C=C/C[C@H]([C@@H](O)[C@@]1(N(C)C([C@]([H])(C(C)C)N(C)C([C@H](CC(C)C)N(C)C([C@]([H])(CC(C)C)N(C)C([C@@H](C)NC([C@H](C)NC([C@@H](N(C([C@@]([H])(NC([C@@H](N(C(CN(C([C@@H](NC1=O)CC)=O)C)=O)C)CC(C)C)=O)C(C)C)=O)C)CC(C)C)=O)=O)=O)=O)=O)=O)[H])C | |
1202.61 | |
Research |
Safety and Handling
H302∣H350∣H360
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.