missing translation for 'onlineSavingsMsg'
Learn More
Cyclosporin A, MedChemExpress Product Code.: 16462176

Cyclosporin A, MedChemExpress

Product Code. 16462176
500 mg, 500mg
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16462176

Brand: Medchem Express HYB0579

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyclosporin A (Cyclosporine A) is an immunosuppressant which binds to the cyclophilin and inhibits phosphatase activity of calcineurin with an IC50 of 5 nM. Cyclosporin A also inhibits CD11a/CD18 adhesion.

Cyclosporin A (Cyclosporine A) is an immunosuppressant which binds to the cyclophilin and inhibits phosphatase activity of calcineurin with an IC50 of 5 nM. Cyclosporin A also inhibits CD11a/CD18 adhesion.

Specifications

Chemical Name or Material Cyclosporin A
Physical Form Solid
CAS 59865-13-3
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C62H111N11O12
For Use With (Application) COVID-19-immunoregulation
Synonym Cyclosporine A Ciclosporin A CsA
Solubility Information DMSO : 62.5 mg/mL (51.97 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (41.58 mM; Need ultrasonic)
SMILES C/C=C/C[C@H]([C@@H](O)[C@@]1(N(C)C([C@]([H])(C(C)C)N(C)C([C@H](CC(C)C)N(C)C([C@]([H])(CC(C)C)N(C)C([C@@H](C)NC([C@H](C)NC([C@@H](N(C([C@@]([H])(NC([C@@H](N(C(CN(C([C@@H](NC1=O)CC)=O)C)=O)C)CC(C)C)=O)C(C)C)=O)C)CC(C)C)=O)=O)=O)=O)=O)=O)[H])C
Molecular Weight (g/mol) 1202.61
Formula Weight 1202.61
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Cyclosporin A, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.