Learn More
SHOWA Cut and Abrasion-Resistant Knit Gloves
The 542 cut and abrasion-resistant glove safeguards your hands while working with components or in hazardous situations. Its polyurethane coating over a reinforced HPPE liner shields against abrasion and oils while allowing for maximum flexibility and dexterity.
1025.00 DKK - 1050.00 DKK
Specifications
Construction | Knit |
---|---|
Coating Material | Polyurethane |
Color | White |
Hand | Specific |
Latex Free | Yes |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
17825532
|
SHOWA
542-06S |
Small |
1025.00 DKK
10 pairs |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17805532
|
SHOWA
542-07M |
Medium |
1025.00 DKK
10 pairs |
|
||||||
17895522
|
SHOWA
542-08L |
Large |
1025.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17835532
|
SHOWA
542-09XL |
X-Large |
1050.00 DKK
10 pairs |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
17815532
|
SHOWA
542-10XXL |
2X-Large |
1025.00 DKK
10 pairs |
Estimated Shipment: 24-07-2025
Log in to see stock. |
||||||
Description
- The SHOWA 542 cut and abrasion-resistant glove is a high-quality protective glove that is designed to keep your hands safe during hazardous applications and component assembly tasks.
- This glove features a unique combination of materials that provide superior protection against cuts, abrasion, and oils, while still maintaining excellent flexibility and dexterity.
- The polyurethane coating enhances the glove's grip, allowing you to work with small components or slippery materials with ease. Additionally, the HPPE reinforced liner provides excellent cut resistance, making this glove ideal for handling sharp objects.
- Designed with comfort in mind. The open back of hand design allows air to circulate around your hands, reducing perspiration and keeping your hands cool and comfortable. The seamless liner minimizes irritation and friction, even during extended wear periods, so you can keep your hands protected without sacrificing comfort.
- The unique combination of materials allows for maximum flexibility, so you can easily handle small components or perform intricate tasks without feeling restricted.
- The SHOWA 542 glove is available in a range of sizes, making it easy to find the perfect fit for your hands. Whether you're working in an industrial setting, handling sharp objects, or working with hazardous materials, the SHOWA 542 glove provides the protection and comfort you need to get the job done safely and efficiently.
Specifications
Knit | |
White | |
Yes | |
Cut- and Abrasion-resistant Glove | |
Unisex | |
ASTM F2878 LEVEL 3 |
Polyurethane | |
Specific | |
Polyurethane | |
EN388:4341B | |
Elasticated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.