Få mere at vide
(±)-Cryptone, TRC
Artikelnummer. 30133856
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133856 500 mg
30133134 250 mg
30008130 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133856 Leverandør Toronto Research Chemicals Leverandørnr. TRCC827465500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 500-02-7
Molekylær formel C9H14O
Molekylvægt (g/mol) 138.21
Synonym 4-(1-Methylethyl)-2-cyclohexen-1-one, 4-Isopropyl-2-cyclohexen-1-one, 4-Isopropyl-2-cyclohexenone, Crypton, DL-Kryptone, NSC 22060
IUPAC navn 4-propan-2-ylcyclohex-2-en-1-one
SMIL CC(C(C=C1)CCC1=O)C
Tekniske data
Kemisk navn eller materiale (±)-Cryptone
Renhedsgrad noter GC
Mængde 500 mg
InChI formel InChI=1S/C9H14O/c1-7(2)8-3-5-9(10)6-4-8/h3,5,7-8H,4,6H2,1-2H3
Formel vægt 138.1045
Procent renhed >95%