Learn More
Thermo Scientific™ CaptureSelect™ MultiSpecies Albumin Depletion Product
This is one of a group of Thermo Scientific™ CaptureSelect™ Proteomics Toolbox™ affinity resins which are unique affinity resins developed for the depletion of proteins from human plasma or serum samples.
8115.00 DKK - 70965.00 DKK
Specifications
Content And Storage | Store at 2°C to 8°C. |
---|---|
Form | Liquid Suspension |
Product Line | CaptureSelect™ |
Type | Albumin Depletion Product |
Stationary Phase | Anti-albumin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18406778
|
Thermo Scientific™
191085305 |
5 mL |
8115.00 DKK
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18476588
|
Thermo Scientific™
191085310 |
10 mL |
14905.00 DKK
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18446548
|
Thermo Scientific™
191085350 |
50 mL |
70965.00 DKK
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This is one of a group of CaptureSelect Proteomics Toolbox™ affinity resins which are unique affinity resins developed for the depletion of proteins from human plasma or serum samples. These affinity ligands are immobilized on a high capacity agarose support for easy, lower-pressure protein depletion or sample prep applications.
Specifications
Store at 2°C to 8°C. | |
CaptureSelect™ | |
Anti-albumin |
Liquid Suspension | |
Albumin Depletion Product |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.