missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CaptureSelect™ MultiSpecies Albumin Depletion Product
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mL
10 mL
50 mL
Unit Size:
10mL
50mL
5mL
Description
This is one of a group of CaptureSelect Proteomics Toolbox™ affinity resins which are unique affinity resins developed for the depletion of proteins from human plasma or serum samples. These affinity ligands are immobilized on a high capacity agarose support for easy, lower-pressure protein depletion or sample prep applications.
Specifications
Specifications
Content And Storage | Store at 2°C to 8°C. |
Form | Liquid Suspension |
Product Line | CaptureSelect™ |
Quantity | 50 mL |
Type | Albumin Depletion Product |
Stationary Phase | Anti-albumin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ CaptureSelect™ MultiSpecies Albumin Depletion Product >
Spot an opportunity for improvement?Share a Content Correction