Learn More
Motic™ C-Mount Camera Adapter for SMZ168 Stereo Zoom Trinocular Microscope Chip Sensor
Mount a digital C-mount camera with 1/3 in., 1/2 in., or 2/3 in. on board chip on your SMZ168 Trinocular
415.00 DKK - 718.00 DKK
Specifications
Type | Camera Adapter |
---|---|
For Use With (Equipment) | SMZ168 Stereo Zoom Microscope |
Quantity | 1 Ea. |
Product Code | Brand | Magnification Power | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Magnification Power | Diameter (Metric) | Price | Quantity & Availability | ||||
11888743
|
Motic™
SW0113N7 |
0.45X | 8.46 mm |
718.00 DKK
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||||
11844313
|
Motic™
SW0113N6 |
0.5X | 12.7, 16.9 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- For use with SMZ168 Stereo Zoom Trinocular Microscope:
- SW0113N6: Mount a digital C-mount camera with 1/2 in. and 2/3 in. on board chip for 0.5X magnification power
- SW0113N7: Mount a digital C-mount camera with 1/3 in. on board chip for 0.45X magnification power
Specifications
Camera Adapter | |
SMZ168 Stereo Zoom Microscope | |
1 Ea. |