Learn More
Motic™ C-Mount Camera Adapter for SMZ168 Stereo Zoom Trinocular Microscope Chip Sensor
Mount a digital C-mount camera with 1/3 in., 1/2 in., or 2/3 in. on board chip on your SMZ168 Trinocular
Brand:  Motic™ SW0113N7
Additional Details : Weight : 0.04888kg
 View more versions of this product
Product Code. 11888743
  • 718.00 DKK / Each
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • For use with SMZ168 Stereo Zoom Trinocular Microscope:
    • SW0113N6: Mount a digital C-mount camera with 1/2 in. and 2/3 in. on board chip for 0.5X magnification power
    • SW0113N7: Mount a digital C-mount camera with 1/3 in. on board chip for 0.45X magnification power
Specifications
Specifications
Camera Adapter
8.46 mm
0.45X
1/3 in.
SMZ168 Stereo Zoom Microscope
1 Ea.