Learn More
Cadence Science™ Non-Coring Septum Penetration Needles Product Code.: 11766844
Cadence Science™ Non-Coring Septum Penetration Needles
Product Code. 11766844
20 G, 8 in., Pack of 12
Click to view available options
Needle Gauge:
18 G
20 G
Length (English) Needle:
1 in.
6 in.
8 in.
12 in.
Unit Size:
Pack of 12
This item is not returnable. View return policy
Product Code. 11766844
Brand: Cadence Science™ 9864
This item is not returnable. View return policy
Needles allow for easier penetration and minimize coring and tracking
  • Micro-mate 316 stainless-steel hub
  • 316 stainless-steel tubing
Specifications
Material Stainless Steel (Hub)
Needle Gauge 20 G
For Use With (Application) For easier penetration and minimize coring and tracking
Length (English) Needle 8 in.
Product Type Penetration Needle
Quantity 12 Needles
Needle Point Style Deflected End
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cadence Science™ Non-Coring Septum Penetration Needles >