missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 10, Scharlab
Used to fit the pH of the reaction media.
125.00 DKK - 166.00 DKK
Specifications
Melting Point | -6°C |
---|---|
Boiling Point | 110°C |
Density | 1.00g/cm3 |
pH | 10.0 |
Physical Form | Solution |
Formulation | Per liter: 2.64g Sodium carbonate, 2.09g Sodium hydrogen carbonate |
Traceability to NIST | Traceable to NIST |
Solubility Information | Miscible in water (20°C) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11329913
|
Scharlab
SO10100250 |
250 mL |
125.00 DKK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11339913
|
Scharlab
SO10101000 |
1 L |
166.00 DKK
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Standard buffer solutions are prepared using gravimetric and volumetric procedures. The batch value is determined by measurement with a combination glass electrode against five-point calibration according to DIN 19268. This pH buffer solution is traceable to Standard Reference Material from NIST.
Specifications
-6°C | |
1.00g/cm3 | |
Solution | |
Traceable to NIST |
110°C | |
10.0 | |
Per liter: 2.64g Sodium carbonate, 2.09g Sodium hydrogen carbonate | |
Miscible in water (20°C) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution pH 10, Scharlab