Få mere at vide
Bis(4-tert-butylphenyl)amine, TRC
Artikelnummer. 30240907
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30240907 250 mg
30240638 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30240907 Leverandør Toronto Research Chemicals Leverandørnr. TRCB490523250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4627-22-9
Molekylær formel C20H27N
Molekylvægt (g/mol) 281.44
Synonym 4,4'-Di-tert-butyldiphenylamine, Benzenamine, 4-(1,1-dimethylethyl)-N-[4-(1,1-dimethylethyl)phenyl]-, Diphenylamine, 4,4'-di-tert-butyl- (7CI,8CI), 4-(1,1-Dimethylethyl)-N-[4-(1,1-dimethylethyl)phenyl]benzenamine, 4,4'-Di-tert-butyldiphenylamine, Bis(4-tert-butylphenyl)amine, Bis(p-tert-butylphenyl)amine, Di-4-tert-butylphenylamine, N,N-Bis(4-tert-butylphenyl)amine, N,N-Di-(p-tert-butylphenyl)amine, N-(4-tert-Butylphenyl)-4-tert-butylaniline, Stearer STAR, p,p'-Di-tert-butyldiphenylamine, 4,4'-Di-tert-butyldiphenylamine, Benzenamine, 4-(1,1-dimethylethyl)-N-[4-(1,1-dimethylethyl)phenyl]-, Diphenylamine, 4,4'-di-tert-butyl- (7CI,8CI), 4-(1,1-Dimethylethyl)-N-[4-(1,1-dimethylethyl)phenyl]benzenamine, 4,4'-Di-tert-butyldiphenylamine, Bis(4-tert-butylphenyl)amine, Bis(p-tert-butylphenyl)amine, Di-4-tert-butylphenylamine, N,N-Bis(4-tert-butylphenyl)amine, N,N-Di-(p-tert-butylphenyl)amine, N-(4-tert-Butylphenyl)-4-tert-butylaniline, Stearer STAR, p,p'-Di-tert-butyldiphenylamine
IUPAC navn 4-tert-butyl-N-(4-tert-butylphenyl)aniline
SMIL CC(C)(C)c1ccc(Nc2ccc(cc2)C(C)(C)C)cc1
Tekniske data
Kemisk navn eller materiale Bis(4-tert-butylphenyl)amine
Mængde 250 mg
InChI formel InChI=1S/C20H27N/c1-19(2,3)15-7-11-17(12-8-15)21-18-13-9-16(10-14-18)20(4,5)6/h7-14,21H,1-6H3
Formel vægt 281.2143