Learn More
Sperian™ Honeywell™ Bionic Faceshield Visors
Offering maximum protection in an ergonomic design. Sperian™ Honeywell™ Bionic Faceshield Visors are incredibly adjustable, with 2784 possible wearer positions.
118.00 DKK - 725.00 DKK
Product Code | Brand | Description | Lens Material | Lens Coating | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Lens Material | Lens Coating | Price | Quantity & Availability | |||||
12645053
|
Sperian™
1011626 |
Visor only | Acetate | Uncoated |
123.00 DKK
Each |
Estimated Shipment: 19-06-2025
Log in to see stock. |
|||||
12655053
|
Sperian™
1011627 |
Visor only | Polycarbonate | Anti-fog |
290.00 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
|||||
12635083
|
Sperian™
1011933 |
Complete faceshield unit | Acetate | Uncoated |
725.00 DKK
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
11826950
|
Sperian™
1011623 |
Complete faceshield unit | Polycarbonate | Uncoated | N/A | ||||||
12635053
|
Sperian™
1011625 |
Visor only | Polycarbonate | N/A | |||||||
Description
- Extended top of head and chin protection
- Large superior-quality visors for maximum visibility
- Available with either polycarbonate visor for impact resistance or acetate visor for chemical protection
- Comfortable fit with goggles or respirators
- Locking mechanism for more complete safety
- Dielectric design
- Fully adjustable ratchet headgear and headband
- Multiple tilt visor positions
- Extra-soft cushioning on all parts of the shield in contact with the head
- Breathable extra-soft sweatband is removable for easy washing and replacement
- Excellent for the agriculture, chemical, forestry, steels and metals and wind energy industries
- Protects against abrasion, airborne particles, biohazards, chemicals, impact, splashes and more
Specifications
Clear | |
Faceshield |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sperian™ Honeywell™ Bionic Faceshield Visors