Få mere at vide
Benzyl-d7 Alcohol, TRC
Artikelnummer. 30179562
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179562 50 mg
30178996 100 mg
30179290 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179562 Leverandør Toronto Research Chemicals Leverandørnr. TRCT53664850MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 71258-23-6
Molekylær formel C7 D7 H O
Molekylvægt (g/mol) 115.181
Synonym Benzene-2,3,4,5,6-d5-methan-α,α-d2-ol (ACI), Benzene-d5-methan-d2-ol (9CI), Benzyl alcohol-d7, Benzyl-d7 alcohol, Benzyl Alcohol-D7
IUPAC navn dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methanol
SMIL [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])O
Tekniske data
Kemisk navn eller materiale Benzyl-d7 Alcohol
Mængde 50 mg
InChI formel InChI=1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2/i1D,2D,3D,4D,5D,6D2
Formel vægt 115.101