CAS RN 71258-23-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
Benzyl-d7 Alcohol, TRC
CAS: 71258-23-6 Molekylær formel: C7 D7 H O Molekylvægt (g/mol): 115.181 Synonym: Benzene-2,3,4,5,6-d5-methan-α,α-d2-ol (ACI),Benzene-d5-methan-d2-ol (9CI),Benzyl alcohol-d7,Benzyl-d7 alcohol,Benzyl Alcohol-D7 IUPAC navn: dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methanol SMIL: [2H]c1c([2H])c([2H])c(c([2H])c1[2H])C([2H])([2H])O
2
Benzyl-d7 Alcohol, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.