Learn More
Benzyl carbamate, 98%
CAS: 621-84-1 | C8H9NO2 | 151.17 g/mol
279.00 DKK - 722.00 DKK
Chemical Identifiers
CAS | 621-84-1 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.17 |
MDL Number | MFCD00007965 |
InChI Key | PUJDIJCNWFYVJX-UHFFFAOYSA-N |
Synonym | carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421 |
PubChem CID | 12136 |
IUPAC Name | benzyl carbamate |
SMILES | NC(=O)OCC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10172320
|
Thermo Scientific Acros
105890250 |
25 g | Glass Bottle |
279.00 DKK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10664942
|
Thermo Scientific Acros
105891000 |
100 g | Glass Bottle |
722.00 DKK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
621-84-1 | |
151.17 | |
PUJDIJCNWFYVJX-UHFFFAOYSA-N | |
12136 | |
NC(=O)OCC1=CC=CC=C1 |
C8H9NO2 | |
MFCD00007965 | |
carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421 | |
benzyl carbamate |
Spezifikation
86.0°C to 89.0°C | |
97.5 | |
White to Beige | |
97.5% min. (GC) | |
H2NCO2CH2C6H5 | |
06, 437 | |
carbamic acid benzyl ester, carbamic acid, phenylmethyl ester, o-benzyl carbamate, ccris 5070, phenylmethyl aminooate, carbamic acid, benzyl ester, benzyl-carbamate, benzyloxycarbonylamine, cbznh2, pubchem12421 | |
NC(=O)OCC1=CC=CC=C1 | |
151.17 | |
151.16 | |
Powder or Flakes |
621-84-1 | |
100.0 | |
Authentic | |
C8H9NO2 | |
MFCD00007965 | |
13,79 | |
PUJDIJCNWFYVJX-UHFFFAOYSA-N | |
benzyl carbamate | |
12136 | |
98% | |
Benzyl carbamate, 99% |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 210-710-4
- Benzyl carbamate
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.